Tripropylamine 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A10285-AK
A10285-K2
CAAAAA10285-AKEA
50.72
CAD
CAAAAA10285-AK
CAAAAA10285-K2
Tripropylamine 98%
Tripropylamine
Purity: 98%
CAS No.: 102-69-2
Molecular Formula: C9H21N
Formula Weight: 143.27
Flash Point: 34°(93°F)
Refractive Index: 1.4160
Odor: Amine-like
Melting Point: -93°
Boiling Point: 156°
UN No.: 2260
MDL No.: MFCD00009363
Formula:
C₉H₂₁N Boiling Pt: 156 °C Melting Pt: –93 °C Density: 0.755 Flash Pt: 34 °C (93 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009363 CAS Number: 102-69-2 EINECS: 203-047-7 UN: 2260 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...