Lead(II) sulfide Pb ≥ 82% (≥82% Pb)

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Lead sulfide, Lead sulphide

A10268-0E A10268-36
CAAAAA10268-0EEA 431.34 CAD
CAAAAA10268-0E CAAAAA10268-36
Lead(II) sulfide Pb ≥ 82% (≥82% Pb)
Lead (II) sulphide

CAS No.: 1314-87-0 Molecular Formula: PbS Formula Weight: 239.25 Refractive Index: 3.921 Odor: Odorless Melting Point: 1114° UN No.: 3077 MDL No.: MFCD00016280

Formula: ̍PbS
MW: 239.27 g/mol
Boiling Pt: 1344
Melting Pt: 1114 °C
Density: 7.5
Storage Temperature: Ambient
MDL Number: MFCD00016280
CAS Number: 1314-87-0
EINECS: 215-246-6
UN: 3077
ADR: 9 III
Merck Index: 14,5421

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR