Tetrabutylammonium bromide 98+%

Supplier: Thermo Scientific Chemicals
Warning

A10249-0E A10249-22 A10249-36
CAAAAA10249-0EEA 1077.56 CAD
CAAAAA10249-0E CAAAAA10249-22 CAAAAA10249-36
Tetrabutylammonium bromide 98+%
Tetrabutylammonium bromide

Phase transfer catalyst


Purity: 98+% CAS No.: 1643-19-2 Molecular Formula: C16H36BrN Formula Weight: 322.38 Odor: Amine-like Melting Point: 101-105° Beilstein Reference: 3570983 MDL No.: MFCD00011633

Formula: C₁₆H₃₆BrN
MW: 322.37 g/mol
Melting Pt: 101…105 °C
Density: 1.15
Storage Temperature: Ambient
MDL Number: MFCD00011633
CAS Number: 1643-19-2
EINECS: 216-699-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR