Anthracene 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A10203-14
A10203-22
A10203-36
CAAAAA10203-14EA
98.91
CAD
CAAAAA10203-14
CAAAAA10203-22
CAAAAA10203-36
Anthracene 99%
Anthracene
Purity: 99%
CAS No.: 120-12-7
Molecular Formula: C14H10
Formula Weight: 178.23
Flash Point: 121°(249°F)
Melting Point: 214-218°
Boiling Point: 340-342°
Beilstein Reference: 1905429
Merck Reference: 14,682
UN No.: 3077
MDL No.: MFCD00001240
Formula:
C₁₄H₁₀ Boiling Pt: 340…342 °C Melting Pt: 214…218 °C Density: 1.250 Flash Pt: 121 °C (249 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00001240 CAS Number: 120-12-7 EINECS: 204-371-1 UN: 3077 ADR: 9,III Merck Index: 14,682 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...