Trifluoromethanesulfonic acid 98+%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Trifluoromethanesulphonic acid, Trifluoromethanesulfonic acid

A10173-09 A10173-30
CAAAAA10173-09EA 56.88 CAD
CAAAAA10173-09 CAAAAA10173-18 CAAAAA10173-30
Trifluoromethanesulfonic acid 98+%
Trifluoromethanesulphonic acid

Purity: 98+% CAS No.: 1493-13-6 Molecular Formula: CHF3O3S Formula Weight: 150.07 Flash Point: None Refractive Index: 1.3270 Odor: Pungent Melting Point: -40° Boiling Point: 161-162° Beilstein Reference: 1812100 Merck Reference: 14,9676 UN No.: 3265 MDL No.: MFCD00007514

Formula: CHF₃O₃S
MW: 150.08 g/mol
Boiling Pt: 162
Melting Pt: 35
Density: 1.708
Storage Temperature: Ambient
MDL Number: MFCD00007514
CAS Number: 1493-13-6
EINECS: 216-087-5
UN: 3265
ADR: 8 III
Merck Index: 14,9676

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR