Cadmium(II) sulfate hydrate 98%

Supplier: Thermo Scientific Chemicals
Danger

A10168-0E A10168-22 A10168-36
CAAAAA10168-0EEA 872.16 CAD
CAAAAA10168-0E CAAAAA10168-22 CAAAAA10168-36
Cadmium(II) sulfate hydrate 98%
Cadmium (II) sulphate hydrate

Purity: 98% CAS No.: 15244-35-6 Molecular Formula: 3CdSO4.xH2O (x≈1) Formula Weight: 208.46(anhy) Refractive Index: 1.565 Odor: Odorless Merck Reference: 14,1627 UN No.: 2570 MDL No.: MFCD00151655

Formula: CdO₄S·H₂O
MW: 226.49 g/mol
Melting Pt: 41.5 °C trans pt
Density: 3.08
Storage Temperature: Ambient
MDL Number: MFCD00151655
CAS Number: 15244-35-6
EINECS: 233-331-6
UN: 2570
ADR: 6.1 III
Merck Index: 14,1627

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR