Ethyl salicylate 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A10101-22
A10101-36
CAAAAA10101-0EEA
336.11
CAD
CAAAAA10101-0E
CAAAAA10101-22
CAAAAA10101-36
Ethyl salicylate 99%
Ethyl salicylate
Purity: 99%
CAS No.: 118-61-6
Molecular Formula: C9H10O3
Formula Weight: 166.18
Flash Point: 107°(224°F)
Refractive Index: 1.5220
Melting Point: 1°
Boiling Point: 232°
Beilstein Reference: 907659
Merck Reference: 14,3850
MDL No.: MFCD00002215
Formula:
C₉H₁₀O₃ Boiling Pt: 232 °C Melting Pt: 1 °C Density: 1.130 Flash Pt: 107 °C(224 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00002215 CAS Number: 118-61-6 EINECS: 204-265-5 Merck Index: 14,3850 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...