Gibberellic acid 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
45569-04
45569-09
CAAA45569-04EA
147.41
CAD
CAAA45569-04
CAAA45569-09
Gibberellic acid 98%
Gibberellic acid
Plant growth hormone
Purity: 98%
CAS No.: 77-06-5
Molecular Formula: C19H22O6
Formula Weight: 346.37
Physical Form: Powder
Appearance: White to off-white
Melting Point: 222° dec.
Beilstein Reference: 54346
Merck Reference: 14,4419
Optical Rotation: +78° (c=2 in methanol)
MDL No.: MFCD00079329
Plant growth hormone
Formula:
C₁₉H₂₂O₆ Melting Pt: 222 °C (dec.) Density: 1.386 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00079329 CAS Number: 77-06-5 EINECS: 201-001-0 Merck Index: 14,4419 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...