Zinc oxide, NanoTek®

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Zinci oxidum, Zinc (II) oxide

44899-22 44899-36
CAAA44899-22EA 59.72 CAD
CAAA44899-22 CAAA44899-36
Zinc oxide, NanoTek®
Zinc oxide

NanoTek® zinc oxide, due to the small particle size, can provide UV protection while maintaining a high level of transparency in coatings, polymers, caulks, adhesives and other resin systems


CAS No.: 1314-13-2 Molecular Formula: ZnO Formula Weight: 81.37 Physical Form: 40-100nm APS Powder, S.A. 10-25m2/g Density: 5.6 Odor: Odorless Merck Reference: 14,10147 UN No.: 3077 MDL No.: MFCD00011300


NanoTek® zinc oxide, due to the small particle size, can provide UV protection while maintaining a high level of transparency in coatings, polymers, caulks, adhesives and other resin systems

Formula: ZnO
MW: 81.39 g/mol
Melting Pt: 1975 °C
Density: 5.6
Storage Temperature: Ambient
MDL Number: MFCD00011300
CAS Number: 1314-13-2
EINECS: 215-222-5
UN: 3077
ADR: 9 III
REACH: 01-2119463881-32
Merck Index: 14,10147

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR