Zirconium(IV) oxide ≥99% (metals basis excluding Hf), nanopowder
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Zirconium dioxide, Zirconium Oxide
44886-14
44886-22
44886-36
CAAAA44886-14EA
142.52
CAD
CAAAA44886-14
CAAAA44886-22
CAAAA44886-36
Zirconium(IV) oxide ≥99% (metals basis excluding Hf), nanopowder
Zirconium (IV) oxide
Purity: 99%
CAS No.: 1314-23-4
Molecular Formula: O2Zr
Formula Weight: 123.22
Physical Form: 15-25nm APS Powder, S.A. 20-30 m2/g
Density: 5.89
Appearance: White
Odor: Odorless
Melting Point: ca 2700°
Boiling Point: ca 5000°
Merck Reference: 14,10180
MDL No.: MFCD00011310
Formula:
ZrO₂ MW: 123.22 g/mol Boiling Pt: 4300 Melting Pt: 2680 Density: 5.89 Storage Temperature: Ambient |
MDL Number:
MFCD00011310 CAS Number: 1314-23-4 EINECS: 215-227-2 Merck Index: 14,10180 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...