Iodine, ultra dry ≥99.999% (metals basis), lumps

Supplier: Thermo Scientific Chemicals
Warning

44857-06 44857-14 44857-22
CAAAA44857-06EA 74.58 CAD
CAAAA44857-06 CAAAA44857-14 CAAAA44857-22
Iodine, ultra dry ≥99.999% (metals basis), lumps

Purity: 99.999% CAS No.: 7553-56-2 Molecular Formula: I2 Formula Weight: 253.81 Physical Form: 2-8mm Typically, Ampouled under argon Density: 4.930 Melting Point: 183-186° subl. UN No.: 3495 MDL No.: MFCD00011355

Formula: I₂
Boiling Pt: 184.3 °C (1013 hPa)
Melting Pt: 183…186 °C (subl.)
Density: 4.930
Storage Temperature: Ambient
MDL Number: MFCD00011355
CAS Number: 7553-56-2
EINECS: 231-442-4
UN: 3495
ADR: 8,III
Merck Index: 13,05036

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR