Tricyclohexylphosphine, Technical Grade
Supplier: Thermo Scientific Chemicals
|
Warning
|
44854-03
44854-06
CAAAA44854-03EA
42.34
CAD
CAAAA44854-03
CAAAA44854-06
Tricyclohexylphosphine, Technical Grade
Tricyclohexylphosphine
CAS No.: 2622-14-2
Molecular Formula: (C6H11)3P
Formula Weight: 280.42
Physical Form: Solid
Sensitivity: Air Sensitive
Appearance: White to off-white
Odor: Unpleasant
Melting Point: 76-78°
MDL No.: MFCD00003853
Formula:
C₁₈H₃₃P Boiling Pt: 132…134 °C (0.02 mmHg) Melting Pt: 76…78 °C Density: 0.9 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003853 CAS Number: 2622-14-2 EINECS: 220-069-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...