Disodium malonate 99%

Supplier: Thermo Scientific Chemicals

44841-14 44841-22
CAAAA44841-14EA 62.09 CAD
CAAAA44841-14 CAAAA44841-22
Disodium malonate 99%
Disodium malonate

Purity: 99% CAS No.: 141-95-7 Molecular Formula: CH2(COONa)2 Formula Weight: 148.03 Physical Form: Powder

Formula: C₃H₂Na₂O₄
Storage Temperature: Ambient
MDL Number: MFCD00002708
CAS Number: 141-95-7
EINECS: 205-514-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR