Indium(III) sulfide ≥99.995% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Warning
|
44836-04
44836-09
44836-18
CAAAA44836-04EA
115.66
CAD
CAAAA44836-04
CAAAA44836-09
CAAAA44836-18
Indium(III) sulfide ≥99.995% (metals basis)
Indium (III) sulphide
Purity: 99.995%
CAS No.: 12030-24-9
Molecular Formula: In2S3
Formula Weight: 325.83
Physical Form: Powder
Sensitivity: Moisture Sensitive
Appearance: Red-orangw
Odor: Like rotten eggs
Melting Point: 1050°
MDL No.: MFCD00011061
Formula:
In₂S₃ Melting Pt: 1050 °C Density: 4.89 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011061 CAS Number: 12030-24-9 EINECS: 234-742-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...