Indium(III) sulfide ≥99.98% (metals basis)

Supplier: Thermo Scientific Chemicals
Warning

44835-04 44835-09 44835-18
CAAAA44835-04EA 66.99 CAD
CAAAA44835-04 CAAAA44835-09 CAAAA44835-18
Indium(III) sulfide ≥99.98% (metals basis)
Indium (III) sulphide

Purity: 99.98% CAS No.: 12030-24-9 Molecular Formula: In2S3 Formula Weight: 325.83 Physical Form: Powder Sensitivity: Moisture Sensitive MDL No.: MFCD00011061

Formula: In₂S₃
Melting Pt: 1050 °C
Density: 4,89 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011061
CAS Number: 12030-24-9
EINECS: 234-742-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR