Mercury(II) bromide ACS
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Mercuric bromide, Mercury dibromide
44816-14
44816-22
CAAAA44816-14EA
76.79
CAD
CAAAA44816-14
CAAAA44816-22
Mercury(II) bromide ACS
Mercury (II) bromide
MDL: MFCD00011040
Soluble in hot alcohol, methanol, HCl2 HBr
CAS No.: 7789-47-1
Molecular Formula: Br2Hg
Formula Weight: 360.40
Physical Form: Powder
Density: 6.05
Sensitivity: Light Sensitive
Appearance: White
Odor: Odorless
Melting Point: 236°
Boiling Point: 322-325° subl.
Solubility: Soluble in hot alcohol, methanol, HCl, HBr
Merck Reference: 14,5875
UN No.: 1634
MDL No.: MFCD00011040
Formula:
Br₂Hg Boiling Pt: 322…325 °C (subl.) Melting Pt: 236 °C Density: 6.05 Storage Temperature: Ambient |
MDL Number:
MFCD00011040 CAS Number: 7789-47-1 EINECS: 232-169-3 UN: 1634 ADR: 6.1,II Merck Index: 14,5875 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...