Cyclopentane for HPLC

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Pentamethylene

44814-AK 44814-K2 44814-K7
CAAAA44814-AKEA 83.27 CAD
CAAAA44814-AK CAAAA44814-K2 CAAAA44814-K7
Cyclopentane for HPLC
Cyclopentane

For HPLC and Spectrophotometry


CAS No.: 287-92-3 Molecular Formula: C5H10 Formula Weight: 70.14 Flash Point: -37°(-34°F) Physical Form: Liquid Density: 0.748 Refractive Index: 1.4060 Appearance: Colorless Odor: Petrol-like Melting Point: -95° Boiling Point: 47-49° Beilstein Reference: 1900195 Merck Reference: 14,2741 UN No.: 1146 MDL No.: MFCD00001356


For HPLC and Spectrophotometry

Formula: C₅H₁₀
Boiling Pt: 47…49 °C
Melting Pt: –95 °C
Density: 0.748
Flash Pt: –37 °C (–34 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001356
CAS Number: 287-92-3
EINECS: 206-016-6
UN: 1146
ADR: 3,II
Merck Index: 14,2741
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4dd230057521cc77c90840c7adfece9c74c0a3b1&vid=352021851b7701a448c5b11d43e94d925d03f802&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR