Fullerene nanotube multi-walled ≤8 nm OD, 2-5 nm ID, Length: 0.5-2 µm

Supplier: Thermo Scientific Chemicals
Warning

44790-01 44790-03 44790-06
CAAAA44790-01EA 69.99 CAD
CAAAA44790-01 CAAAA44790-03 CAAAA44790-06
Fullerene nanotube multi-walled ≤8 nm OD, 2-5 nm ID, Length: 0.5-2 µm
Fullerene nanotube

CAS No.: 308068-56-6 Physical Form: Powder UN No.: 1325 MDL No.: MFCD00151408

Melting Pt: > 280 °C
Density: ∼1.65 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00133992
CAS Number: 308068-56-6
UN: 1325
ADR: 4.1,III
Merck Index: 13,01447

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR