Soda lime, with indicator

Supplier: Thermo Scientific Chemicals
Danger

44786-36 44786-A4
CAAAA44786-36EA 261 CAD
CAAAA44786-36 CAAAA44786-A4
Soda lime, with indicator
Soda lime

CAS No.: 8006-28-8 Physical Form: 8-12 Mesh Sensitivity: Air Sensitive and Hygroscopic Odor: Odorless Merck Reference: 14,8569 UN No.: 1907 MDL No.: MFCD00134094

Storage Temperature: Ambient
MDL Number: MFCD00134094
CAS Number: 8006-28-8
UN: 1907
ADR: 8,III
Merck Index: 14,8569

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR