Silicon(IV) oxide, amorphous ≥99.9% (metals basis), nanopowder

Supplier: Thermo Scientific Chemicals
Warning

44781-14 44781-22 44781-36
CAAAA44781-14EA 105.07 CAD
CAAAA44781-14 CAAAA44781-22 CAAAA44781-36
Silicon(IV) oxide, amorphous ≥99.9% (metals basis), nanopowder
Silica gel amorphous

Purity: 99.9% CAS No.: 7631-86-9 Molecular Formula: SiO2 Formula Weight: 60.09 Physical Form: 80nm APS Powder Density: 2.4 Appearance: White Odor: Odorless pH: 6 Melting Point: 1710° Boiling Point: 2230° Merck Reference: 14,8493 MDL No.: MFCD00011232

Formula: SiO₂
Boiling Pt: 2230 °C
Melting Pt: 1710 °C
Density: 2.4
Storage Temperature: Ambient
MDL Number: MFCD00011232
CAS Number: 7631-86-9
EINECS: 231-545-4
Merck Index: 14,8493

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR