Calcium oxide ≥99.95% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Danger
|
44776-18
44776-30
44776-A1
CAAAA44776-18EA
132.72
CAD
CAAAA44776-18
CAAAA44776-30
CAAAA44776-A1
Calcium oxide ≥99.95% (metals basis)
Calcium oxide
Purity: 99.95%
CAS No.: 1305-78-8
Molecular Formula: CaO
Formula Weight: 56.08
Physical Form: -20 Mesh Powder
Density: 3.37
Refractive Index: 1.83
Sensitivity: Air and Moisture Sensitive
Appearance: Off-white
Odor: Odorless
Melting Point: 2572°
Boiling Point: 2850°
Merck Reference: 14,1686
MDL No.: MFCD00010911
Formula:
CaO Boiling Pt: 2850 °C Melting Pt: 2572 °C Density: 3.37 Storage Temperature: Ambient |
MDL Number:
MFCD00010911 CAS Number: 1305-78-8 EINECS: 215-138-9 UN: 1910 ADR: 8,III Merck Index: 14,1686 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...