Lithium chloride, ultra dry ≥99.995% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Warning
|
44762-06
44762-14
44762-22
CAAAA44762-06EA
99.86
CAD
CAAAA44762-06
CAAAA44762-14
CAAAA44762-22
Lithium chloride, ultra dry ≥99.995% (metals basis)
Lithium chloride
Purity: 99.995%
CAS No.: 7447-41-8
Molecular Formula: LiCl
Formula Weight: 42.39
Physical Form: Powder, Ampouled under argon
Density: 2.068
Melting Point: 605°
Boiling Point: 1325-1360°
Merck Reference: 14,5528
MDL No.: MFCD00011078
Formula:
LiCl MW: 42.39 g/mol Boiling Pt: 1325…1360 °C Melting Pt: 605 °C Density: 2.068 Storage Temperature: Ambient |
MDL Number:
MFCD00011078 CAS Number: 7447-41-8 EINECS: 231-212-3 Merck Index: 14,5528 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...