Magnesium oxide ≥99% (metals basis), nanopowder

Supplier: Thermo Scientific Chemicals

Synonyms: Magnesia usta

44733-14 44733-22 44733-36
CAAAA44733-14EA 103.65 CAD
CAAAA44733-14 CAAAA44733-22 CAAAA44733-36
Magnesium oxide ≥99% (metals basis), nanopowder
Magnesium oxide

Purity: 99+% CAS No.: 1309-48-4 Molecular Formula: MgO Formula Weight: 40.30 Physical Form: 100nm APS Powder, S.A. 7.3m2/g Density: 3.58 Refractive Index: 1.736 Odor: Odorless Melting Point: 2852° Boiling Point: 3600° Merck Reference: 14,5677 MDL No.: MFCD00011109

Formula: MgO
MW: 40.3 g/mol
Boiling Pt: 3600 °C
Melting Pt: 2852 °C
Density: 3.58
Storage Temperature: Ambient
MDL Number: MFCD00011109
CAS Number: 1309-48-4
EINECS: 215-171-9
Merck Index: 14,5677

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR