Cobalt(II,III) oxide ≥99% (metals basis), nanopowder

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Tricobalt tetraoxide, Cobaltic tetraoxide

44661-14 44661-22 44661-36
CAAAA44661-14EA 123.08 CAD
CAAAA44661-14 CAAAA44661-22 CAAAA44661-36
Cobalt(II,III) oxide ≥99% (metals basis), nanopowder
Cobalt (II,III) oxide

Purity: 99% CAS No.: 1308-06-1 Molecular Formula: Co3O4 Formula Weight: 240.80 Physical Form: 50-80nm APS Powder, S.A. 10m2/g Density: 6.11 Appearance: Black Melting Point: 900° dec. Merck Reference: 14,2429 UN No.: 3077 MDL No.: MFCD00010939

Formula: Co₃O₄
MW: 240.8 g/mol
Melting Pt: 900 °C (dec.)
Density: 6.11
Storage Temperature: Ambient
MDL Number: MFCD00010939
CAS Number: 1308-06-1
EINECS: 215-157-2
UN: 3077
ADR: 9,III
Merck Index: 14,2429

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR