Silicon carbide, β-phase, nanopowder
Supplier: Thermo Scientific Chemicals
|
Warning
|
44646-06
44646-14
44646-22
CAAAA44646-06EA
76.95
CAD
CAAAA44646-06
CAAAA44646-14
CAAAA44646-22
Silicon carbide, β-phase, nanopowder
Silicon carbide, β-phase
CAS No.: 409-21-2
Molecular Formula: SiC
Formula Weight: 40.10
Physical Form: 45-55nm APS Powder, S.A. 70-90m2/g
Odor: Odorless
Melting Point: 2700°
MDL No.: MFCD00049531
Formula:
SiC Melting Pt: 2700 °C Storage Temperature: Ambient |
MDL Number:
MFCD00049531 CAS Number: 409-21-2 EINECS: 206-991-8 Merck Index: 14,8492 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...