5-(Chloromethyl)uracil 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
44639-01
44639-03
44639-06
CAAAA44639-01EA
86.11
CAD
CAAAA44639-01
CAAAA44639-03
CAAAA44639-06
5-(Chloromethyl)uracil 97%
5-(Chloromethyl)uracil
Purity: 97%
CAS No.: 3590-48-5
Molecular Formula: C5H5ClN2O2
Formula Weight: 160.56
Physical Form: Powder
Formula:
C₅H₅ClN₂O₂ Storage Temperature: Ambient |
MDL Number:
MFCD00218445 CAS Number: 3590-48-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...