Cesium trifluoroacetate

Supplier: Thermo Scientific Chemicals
Warning

44633-09 44633-18
CAAAA44633-09EA 77.1 CAD
CAAAA44633-09 CAAAA44633-18
Cesium trifluoroacetate
Cesium trifluoroacetate

CAS No.: 21907-50-6 Molecular Formula: CsOOCCF3 Formula Weight: 245.92 Physical Form: Crystalline Melting Point: 115° MDL No.: MFCD00051304

Formula: C₂CsF₃O₂
Melting Pt: 115 °C
Storage Temperature: Ambient
MDL Number: MFCD00051304
CAS Number: 21907-50-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR