Trifluoroacetic acid ≥99.5% for HPLC
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
TFA
44630-AP
44630-AY
CAAAA44630-AEEA
441.52
CAD
CAAAA44630-AE
CAAAA44630-AP
CAAAA44630-5Y
CAAAA44630-AY
Trifluoroacetic acid ≥99.5% for HPLC
Trifluoroacetic acid
Purity: 99.5+%
CAS No.: 76-05-1
Molecular Formula: CF3CO2H
Formula Weight: 114.02
Physical Form: Liquid
Density: 1.480
Refractive Index: 1.2840
Sensitivity: Hygroscopic
Odor: Pungent
Melting Point: -15°
Boiling Point: 72-73°
Beilstein Reference: 742035
Merck Reference: 14,9681
UN No.: 2699
MDL No.: MFCD00004169
Formula:
C₂HF₃O₂ MW: 114.02 g/mol Boiling Pt: 71.8 Melting Pt: -15 Density: 1.480 Storage Temperature: Ambient |
MDL Number:
MFCD00004169 CAS Number: 76-05-1 EINECS: 200-929-3 UN: 2699 ADR: 8 I Merck Index: 14,9681 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...