2-Amino-5-bromo-4-methylpyridine ≥97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
5-Bromo-4-methylpyridin-2-amine, 5-Bromo-4-methyl-2-pyridylamine, 2-Amino-5-bromo-4-picoline
44623-03
44623-06
44623-14
CAAAA44623-03EA
42.82
CAD
CAAAA44623-03
CAAAA44623-06
CAAAA44623-14
2-Amino-5-bromo-4-methylpyridine ≥97%
2-Amino-5-bromo-4-methylpyridine
Purity: 97%
CAS No.: 98198-48-2
Molecular Formula: C6H7BrN2
Formula Weight: 187.04
Physical Form: Powder
Appearance: Cream
Melting Point: 148-151°
MDL No.: MFCD03427660
Formula:
C₆H₇BrN₂ MW: 187.04 g/mol Melting Pt: 148…151 °C Storage Temperature: Ambient |
MDL Number:
MFCD03427660 CAS Number: 98198-48-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...