Praseodymium(III,IV) oxide ≥99% (REO, rare earth oxide basis)
Supplier: Thermo Scientific Chemicals
|
Warning
|
44594-30
44594-A1
CAAAA44594-30EA
224.36
CAD
CAAAA44594-30
CAAAA44594-A1
CAAAA44594-A7
Praseodymium(III,IV) oxide ≥99% (REO, rare earth oxide basis)
Praseodymium (III,IV) oxide
Purity: 99%
CAS No.: 12037-29-5
Molecular Formula: Pr6O11
Formula Weight: 1021.43
Physical Form: Powder
Melting Point: 2500°
MDL No.: MFCD00011178
Formula:
O₁₁Pr₆ MW: 1021.44 g/mol Melting Pt: 2500 °C Storage Temperature: Ambient |
MDL Number:
MFCD00011178 CAS Number: 12037-29-5 EINECS: 234-857-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...