Aluminium sulfate, anhydrous ≥99.99% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Danger
|
44563-06
44563-14
44563-22
44563-36
CAAAA44563-06EA
86.9
CAD
CAAAA44563-06
CAAAA44563-14
CAAAA44563-22
CAAAA44563-36
Aluminium sulfate, anhydrous ≥99.99% (metals basis)
Aluminium sulphate
Purity: 99.99%
CAS No.: 10043-01-3
Molecular Formula: Al2(SO4)3
Formula Weight: 342.15
Physical Form: Powder, Packaged under argon
Density: 2.71
pH: 3.5
Melting Point: 770°
Merck Reference: 14,366
MDL No.: MFCD00003423
Formula:
Al₂O₁₂S₃ Melting Pt: 770 °C Density: 2.71 Storage Temperature: Ambient |
MDL Number:
MFCD00003423 CAS Number: 10043-01-3 EINECS: 233-135-0 Merck Index: 14,366 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...