Lanthanum(III) iodide, ultra dry ≥99.995% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Warning
|
44548-03
44548-06
CAAAA44548-03EA
240.16
CAD
CAAAA44548-03
CAAAA44548-06
Lanthanum(III) iodide, ultra dry ≥99.995% (metals basis)
Lanthanum (III) iodide
Purity: 99.995+%
CAS No.: 13813-22-4
Molecular Formula: LaI3
Formula Weight: 519.62
Physical Form: -10 Mesh Beads, Ampouled under argon
Sensitivity: Moisture Sensitive
MDL No.: MFCD00049470
Formula:
I₃La Melting Pt: 772 °C Density: 5.6 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00049470 CAS Number: 13813-22-4 EINECS: 237-465-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...