Europium ≥99.9% (metals basis), rods, Ø 9.52 mm (0.375 in)
Supplier: Thermo Scientific Chemicals
|
Danger
|
44545-DJ
44545-HF
CAAAA44545-DJEA
2753.94
CAD
CAAAA44545-DJ
CAAAA44545-HF
Europium ≥99.9% (metals basis), rods, Ø 9.52 mm (0.375 in)
Europium
Purity: 99.9%
CAS No.: 7440-53-1
Molecular Formula: Packaged in mineral oil
Physical Form: ≈0.37g/mm, Cast rod, diameter approximate
Appearance: Silver/gray
Odor: Odorless
UN No.: 3208
MDL No.: MFCD00010992
Formula:
Eu Storage Temperature: Ambient |
MDL Number:
MFCD00010992 CAS Number: 7440-53-1 EINECS: 231-161-7 UN: 3208 ADR: 4.3,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...