Aluminium acetylacetonate ≥99.995% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Tris(2,4-pentanedionato-O,O')aluminium, Acetylacetone Aluminum(III) Salt, Al(acac)3, Aluminium 2,4-pentanedionate, Aluminum(III) Acetylacetonate, Aluminium tris(2,4-pentanedionato-O,O'), Aluminum acetylacetonate, Aluminum 2,4-pentanedionate, Tris(2,4-pentanedionato)aluminum(III), Aluminum triacetylacetone
Purity: 99.995+%
CAS No.: 13963-57-0
Molecular Formula: C15H21AlO6
Formula Weight: 324.31
Physical Form: Powder
Density: 1.27
Melting Point: 192-194°
Boiling Point: 314-316°
Beilstein Reference: 4157942
UN No.: 3467
MDL No.: MFCD00000013
Formula:
C₁₅H₂₁AlO₆ MW: 324.31 g/mol Boiling Pt: 315 Melting Pt: 192…194 °C Density: 1.27 Storage Temperature: Ambient |
MDL Number:
MFCD00000013 CAS Number: 13963-57-0 EINECS: 237-741-6 UN: 3467 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...