Zinc oxide ≥99% (metals basis), nanopowder

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Zinci oxidum, Zinc (II) oxide

44533-09 44533-18
CAAAA44533-09EA 210.14 CAD
CAAAA44533-09 CAAAA44533-18
Zinc oxide ≥99% (metals basis), nanopowder
Zinc oxide

Purity: 99% CAS No.: 1314-13-2 Molecular Formula: ZnO Formula Weight: 81.37 Physical Form: 20-30nm APS Powder Density: 5.606 Appearance: White Odor: Odorless Melting Point: 1975° Merck Reference: 14,10147 UN No.: 3077 MDL No.: MFCD00011300

Formula: ZnO
MW: 81.39 g/mol
Melting Pt: 1975 °C
Density: 5.606
Storage Temperature: Ambient
MDL Number: MFCD00011300
CAS Number: 1314-13-2
EINECS: 215-222-5
UN: 3077
ADR: 9 III
REACH: 01-2119463881-32
Merck Index: 14,10147

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR