Ethylene glycol 99+% for spectrophotometry
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Ethane-1,2-diol, 1,2-Dihydroxy ethane, 1,2-Ethanediol, Glycol
For use in spectrophotometry
Purity: 99+%
CAS No.: 107-21-1
Molecular Formula: C2H6O2
Formula Weight: 62.07
Flash Point: 119°(246°F)
Physical Form: Liquid
Density: 1.113
Refractive Index: 1.4310
Sensitivity: Hygroscopic
Appearance: Colorless
Odor: Sweet
Melting Point: -13°
Boiling Point: 196-198°
Beilstein Reference: 505945
Merck Reference: 14,3798
MDL No.: MFCD00002885
For use in spectrophotometry
Formula:
HOCH₂CH₂OH MW: 62.07 g/mol Boiling Pt: 196…198 °C Melting Pt: -13 Density: 1.113 Flash Pt: 119 °C (246 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00002885 CAS Number: 107-21-1 EINECS: 203-473-3 REACH: 01-2119456816-28 Merck Index: 14,3798 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...