n-Heptadecane ≥99.5%
Supplier: Thermo Scientific Chemicals
|
Warning
|
44514-03
44514-06
CAAAA44514-03EA
36.97
CAD
CAAAA44514-03
CAAAA44514-06
n-Heptadecane ≥99.5%
n-Heptadecane
Purity: 99.5%
CAS No.: 629-78-7
Molecular Formula: CH3(CH2)15CH3
Formula Weight: 240.48
Physical Form: Crystalline
Density: 0.7783
Refractive Index: 1.4369
Melting Point: 22°
Boiling Point: 302°
MDL No.: MFCD00009002
Formula:
H₃C(CH₂)₁₅CH₃ Boiling Pt: 302 °C Melting Pt: 22 °C Density: 0,7783 Flash Pt: 148 °C Storage Temperature: Ambient |
MDL Number:
MFCD00009002 CAS Number: 629-78-7 EINECS: 211-108-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...