n-Hexadecane ≥99.5%
Supplier: Thermo Scientific Chemicals
|
Danger
|
44511-06
44511-14
CAAAA44511-06EA
100.8
CAD
CAAAA44511-06
CAAAA44511-14
n-Hexadecane ≥99.5%
n-Hexadecane
Purity: 99.5+%
CAS No.: 544-76-3
Molecular Formula: CH3(CH2)14CH3
Formula Weight: 226.45
Flash Point: 135°(275°F)
Physical Form: Liquid
Density: 0.773
Refractive Index: 1.4345
Appearance: Colorless
Melting Point: 18°
Boiling Point: 287°
Beilstein Reference: 1736592
MDL No.: MFCD00008998
Formula:
H₃C(CH₂)₁₄CH₃ Boiling Pt: 287 °C Melting Pt: 18 °C Density: 0.773 Flash Pt: 135 °C (275 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008998 CAS Number: 544-76-3 EINECS: 208-878-9 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...