Amidosulfonic acid ≥99.99% (metals basis)

Supplier: Thermo Scientific Chemicals
Warning

44510-14 44510-22 44510-36
CAAAA44510-14EA 43.92 CAD
CAAAA44510-14 CAAAA44510-22 CAAAA44510-36
Amidosulfonic acid ≥99.99% (metals basis)
Amidosulphonic acid

Purity: 99.99% CAS No.: 5329-14-6 Molecular Formula: H2NSO3H Formula Weight: 97.09 Physical Form: Crystalline Density: 2.12 pH: 1.2 Melting Point: ca 205° dec. Merck Reference: 14,8921 UN No.: 2967 MDL No.: MFCD00011603

Formula: H₃NO₃S
Boiling Pt: 247 °C (1013 hPa)
Melting Pt: ∼ 205 °C (dec.)
Density: 2.12
Storage Temperature: Ambient
MDL Number: MFCD00011603
CAS Number: 5329-14-6
EINECS: 226-218-8
UN: 2967
ADR: 8,III
Merck Index: 14,8921
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=afa0500786be69d531cdf06f8e67548e6f67b23f&vid=73aa20196fbabf965e4375f2cf3675e2f513ee59&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR