Calcium bromide hydrate ≥99.999% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Warning
|
44497-14
44497-22
CAAAA44497-14EA
218.04
CAD
CAAAA44497-14
CAAAA44497-22
CAAAA44497-36
Calcium bromide hydrate ≥99.999% (metals basis)
Calcium bromide hydrate
Purity: 99.999%
CAS No.: 71626-99-8
Molecular Formula: CaBr2.xH2O
Formula Weight: 199.90(anhy)
Physical Form: Crystalline
Appearance: White
MDL No.: MFCD00149608
Formula:
Br₂Ca·H₂O Density: 2.295 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00149608 CAS Number: 71626-99-8 EINECS: 232-164-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...