Potassium hexafluorosilicate ≥99.999% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Danger
|
44495-09
44495-18
44495-30
CAAAA44495-09EA
116.45
CAD
CAAAA44495-09
CAAAA44495-18
CAAAA44495-30
Potassium hexafluorosilicate ≥99.999% (metals basis)
Potassium hexafluorosilicate
Purity: 99.999%
CAS No.: 16871-90-2
Molecular Formula: F6K2Si
Formula Weight: 220.28
Physical Form: Crystalline
Density: 2.27
Appearance: White
pH: 3.4
Melting Point: dec.
Merck Reference: 14,7638
UN No.: 2655
MDL No.: MFCD00040666
Formula:
F₆K₂Si Melting Pt: Dec. Density: 2.27 Storage Temperature: Ambient |
MDL Number:
MFCD00040666 CAS Number: 16871-90-2 EINECS: 240-896-2 UN: 2655 ADR: 6.1,III Merck Index: 14,7638 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...