Lithium hydroxide monohydrate ≥99.995% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Danger
|
44473-14
44473-22
44473-36
CAAAA44473-14EA
169.06
CAD
CAAAA44473-14
CAAAA44473-22
CAAAA44473-36
Lithium hydroxide monohydrate ≥99.995% (metals basis)
Lithium hydroxide monohydrate
Purity: 99.995%
CAS No.: 1310-66-3
Molecular Formula: LiOH.H2O
Formula Weight: 41.96 (23.95anhy)
Physical Form: Powder
Density: 1.51
Merck Reference: 14,5534
UN No.: 2680
MDL No.: MFCD00149772
Formula:
LiOH·1H₂O Boiling Pt: 920 °C (1013 hPa) Melting Pt: 462 °C Density: 1.51 Storage Temperature: Ambient |
MDL Number:
MFCD00149772 CAS Number: 1310-66-3 EINECS: 215-183-4 UN: 2680 ADR: 8,II Merck Index: 14,5534 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...