Lithium hydroxide, anhydrous ≥99.995% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Danger
|
44471-06
44471-14
44471-22
CAAAA44471-06EA
91.01
CAD
CAAAA44471-06
CAAAA44471-14
CAAAA44471-22
Lithium hydroxide, anhydrous ≥99.995% (metals basis)
Lithium hydroxide
Purity: 99.995%
CAS No.: 1310-65-2
Molecular Formula: LiOH
Formula Weight: 23.95
Physical Form: Powder
Density: 2.54
Appearance: White
Odor: Odorless
pH: 14
Melting Point: 462°
Boiling Point: 924°
Merck Reference: 14,5534
UN No.: 2680
MDL No.: MFCD00011095
Formula:
LiOH Boiling Pt: 924 °C Melting Pt: 462 °C Density: 2.54 Storage Temperature: Ambient |
MDL Number:
MFCD00011095 CAS Number: 1310-65-2 EINECS: 215-183-4 UN: 2680 ADR: 8,II Merck Index: 14,5534 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...