6-(Trifluoromethyl)uracil 97%

Supplier: Thermo Scientific Chemicals

Synonyms: 4-(Trifluoromethyl)uracil, 2,4-Dihydroxy-6-trifluoromethylpyrimidine

44467-03 44467-06
CAAAA44467-03EA 83.11 CAD
CAAAA44467-03 CAAAA44467-06
6-(Trifluoromethyl)uracil 97%
6-(Trifluoromethyl)uracil

Purity: 97% CAS No.: 672-45-7 Molecular Formula: C5H3F3N2O2 Formula Weight: 180.08 Physical Form: Powder

Formula: C₅H₃F₃N₂O₂
MW: 180.09 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01011762
CAS Number: 672-45-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR