Cerium tribromide 99%

Supplier: Thermo Scientific Chemicals
Warning

44447-14 44447-22
CAAAA44447-14EA 262.28 CAD
CAAAA44447-14 CAAAA44447-22
Cerium tribromide 99%
Cerium tribromide

Purity: 99% CAS No.: 14457-87-5 Molecular Formula: CeBr3.xH2O Formula Weight: 379.83(anhy) Physical Form: Crystalline Odor: Odorless MDL No.: MFCD00150206

Formula: CeBr₃
Melting Pt: 732 °C
Storage Temperature: Ambient
MDL Number: MFCD00016004
CAS Number: 14457-87-5
EINECS: 238-447-0
Merck Index: 12,02039

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR