3-Nitrobenzyl alcohol 99+%

Supplier: Thermo Scientific Chemicals

44445-03 44445-06
CAAAA44445-03EA 70.63 CAD
CAAAA44445-03 CAAAA44445-06
3-Nitrobenzyl alcohol 99+%
3-Nitrobenzyl alcohol

Purity: 99+% CAS No.: 619-25-0 Molecular Formula: C7H7NO3 Formula Weight: 153.14 Flash Point: 110°(230°F) Physical Form: Solid Density: 1.300 Refractive Index: 1.5730 Appearance: Yellow Melting Point: 29-33° Boiling Point: 175-180°/3mm Beilstein Reference: 2044769 MDL No.: MFCD00007273

Formula: C₇H₇NO₃
Boiling Pt: 175…180 °C (3 mmHg)
Melting Pt: 29…33 °C
Density: 1.300
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007273
CAS Number: 619-25-0
EINECS: 210-588-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR