Barium oxide ≥99.5% (metals basis excluding Sr) (max. 0.05% Sr)
Supplier: Thermo Scientific Chemicals
|
Warning
|
44440-14
44440-22
CAAAA44440-14EA
396.58
CAD
CAAAA44440-14
CAAAA44440-22
Barium oxide ≥99.5% (metals basis excluding Sr) (max. 0.05% Sr)
Barium oxide
Purity: 99.5%
CAS No.: 1304-28-5
Molecular Formula: BaO
Formula Weight: 153.33
Physical Form: Powder
Density: 5.7
Refractive Index: 1.98
Melting Point: 1923°
Merck Reference: 14,986
UN No.: 1884
MDL No.: MFCD00003453
Formula:
BaO Boiling Pt: ∼2000 °C (1013 hPa) Melting Pt: 1923 °C Density: 5.7 Storage Temperature: Ambient |
MDL Number:
MFCD00003453 CAS Number: 1304-28-5 EINECS: 215-127-9 UN: 1884 ADR: 6.1,III Merck Index: 14,986 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...