Cerium triacetate hydrate ≥99.995% (metals basis)

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Acetic acid cerium (III) salt hydrate, Cerium (III) acetate hydrate

44433-09 44433-18 44433-30
CAAAA44433-09EA 112.02 CAD
CAAAA44433-09 CAAAA44433-18 CAAAA44433-30
Cerium triacetate hydrate ≥99.995% (metals basis)
Cerium triacetate hydrate

Purity: 99.995% CAS No.: 206996-60-3 Molecular Formula: C6H9CeO6.xH2O Formula Weight: 317.26(anhy) Physical Form: Crystalline Appearance: White MDL No.: MFCD00051060

Formula: C₆H₉CeO₆·H₂O
MW: 335.266 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00150533
CAS Number: 206996-60-3
EINECS: 208-654-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR