Cerium tribromide ≥99.998% (metals basis)

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Cerous bromide, Cerium(III) bromide hydrate, Cerium (III) bromide

44432-09 44432-18
CAAAA44432-09EA 118.97 CAD
CAAAA44432-09 CAAAA44432-18
Cerium tribromide ≥99.998% (metals basis)
Cerium tribromide

Purity: 99.998% CAS No.: 14457-87-5 Molecular Formula: CeBr3.xH2O Formula Weight: 379.83(anhy) Physical Form: Crystalline Appearance: Pale orange Odor: Odorless Merck Reference: 14,1995 MDL No.: MFCD00150206

Formula: CeBr₃
Melting Pt: 732 °C
Storage Temperature: Ambient
MDL Number: MFCD00016004
CAS Number: 14457-87-5
EINECS: 238-447-0
Merck Index: 14,1995

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR