Europium(II) bromide ≥99.99% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Europium dibromide
44431-03
44431-06
44431-14
CAAAA44431-03EA
173.8
CAD
CAAAA44431-03
CAAAA44431-06
CAAAA44431-14
Europium(II) bromide ≥99.99% (metals basis)
Europium (II) bromide
Purity: 99.99%
CAS No.: 13780-48-8
Molecular Formula: EuBr2
Formula Weight: 311.77
Physical Form: Powder
Sensitivity: Hygroscopic
MDL No.: MFCD01076548
Formula:
Br₂Eu Storage Temperature: Ambient |
MDL Number:
MFCD01076548 CAS Number: 13780-48-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...